Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2H-1-Benzopyran-2-one, 7-hydroxy-3-(1,2,3-trihydroxy-3-methylbutyl)- | 1291053-38-7 | 1 MG
SDP

Catalog No. 5000420968
Encompass_Preferred

Evodosin A is a coumarins product that can be isolated from the herbs of Evodia lepta.

  • CAS number: 1291053-38-7
  • Molecular formula: C14H16O6
  • Molecular weight: 280.27
  • Appearance: Off-white to light yellow solid
  • Purity: 96.0%
  • Storage conditions: Powder at -20°C for 3 years; in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-20968 Supplier Medchemexpress LLC Supplier No. HYN38841MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Evodosin A is a coumarins product that can be isolated from the herbs of Evodia lepta.

  • CAS number: 1291053-38-7
  • Molecular formula: C14H16O6
  • Molecular weight: 280.27
  • Appearance: Off-white to light yellow solid
  • Purity: 96.0%
  • Storage conditions: Powder at -20°C for 3 years; in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.