Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2H-1-Benzopyran-2-one, 7-methoxy-4-methyl- | 2555-28-4 | 98.4% | 250 MG
SDP

Catalog No. 5000414366
Encompass_Preferred

4-Methylherniarin is a coumarin derivative and fluorescent label with antimicrobial activity against both gram-positive and gram-negative bacterial strains. It shows good activity against B. subtilis and S. sonnei with IC50 values of 11.76 μg/ml and 13.47 μg/ml, respectively.

  • Coumarin derivative and fluorescent label
  • Antimicrobial activity against gram-positive and gram-negative bacterial strains
  • Activity against B. subtilis (IC50: 11.76 μg/ml)
  • Activity against S. sonnei (IC50: 13.47 μg/ml)
  • Appearance: Solid, white to off-white
  • For research use only

Catalog No. 50-004-14366 Supplier Medchemexpress LLC Supplier No. 250MGHYD0128
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methylherniarin is a coumarin derivative and fluorescent label with antimicrobial activity against both gram-positive and gram-negative bacterial strains. It shows good activity against B. subtilis and S. sonnei with IC50 values of 11.76 μg/ml and 13.47 μg/ml, respectively.

  • Coumarin derivative and fluorescent label
  • Antimicrobial activity against gram-positive and gram-negative bacterial strains
  • Activity against B. subtilis (IC50: 11.76 μg/ml)
  • Activity against S. sonnei (IC50: 13.47 μg/ml)
  • Appearance: Solid, white to off-white
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.