Learn More
Medchemexpress LLC 2H-1-benzopyran-3-carboxylic acid, 2-oxo- | 531-81-7 | MFCD00006852 | 99.9% | 190.15 | C10H6O4 | 5 G
SDP
Catalog No. 5000301080
Encompass_Preferred
Coumarin-3-carboxylic acid (2-Oxochromene-3-carboxylic acid) is a coumarin derivative used as an intermediate for coumarin synthesis and as a fluorescent probe for hydroxyl radicals in biochemical research. It is supplied as a white to off-white solid with high analytical purity and DMSO solubility.
  • High purity: 99.93%.
  • White to off-white solid form.
  • Soluble in DMSO: 100 mg/mL; ultrasonic assistance recommended.
  • Useful as a fluorescent probe for hydroxyl radicals (•OH).
  • Suitable as a synthetic intermediate for coumarin derivatives.
  • Reported to form lanthanide complexes with antiproliferative activity.
Catalog No. 50-003-01080 Supplier Medchemexpress LLC Supplier No. HYW0279685G
Coumarin-3-carboxylic acid (2-Oxochromene-3-carboxylic acid) is a coumarin derivative used as an intermediate for coumarin synthesis and as a fluorescent probe for hydroxyl radicals in biochemical research. It is supplied as a white to off-white solid with high analytical purity and DMSO solubility.
  • High purity: 99.93%.
  • White to off-white solid form.
  • Soluble in DMSO: 100 mg/mL; ultrasonic assistance recommended.
  • Useful as a fluorescent probe for hydroxyl radicals (•OH).
  • Suitable as a synthetic intermediate for coumarin derivatives.
  • Reported to form lanthanide complexes with antiproliferative activity.