Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2H-1-benzopyran-7-ol, 3,4-dihydro-3-(4-hydroxyphenyl)-4-(4-methoxyphenyl)-(3R,4S) | 1213777-80-0 | 99.9% | 348.39 g/mol | C22H20O4 | 5 MG
SDP

Catalog No. 5000221019
Encompass_Preferred

Triphen diol is a phenol diol small-molecule research compound (CAS 1213777-80-0) reported to induce apoptosis and inhibit proliferation in pancreatic cancer models. It has molecular formula C22H20O4, molecular weight 348.39 g/mol, and is supplied with high reported purity for use in in vitro and in vivo studies. Solubility and storage guidance is provided to support experimental workflows.

  • Phenol diol small-molecule with reported anticancer activity.
  • Demonstrated apoptosis induction and antiproliferative effects in pancreatic cancer cell lines.
  • High reported purity suitable for biological assays.
  • Soluble in DMSO at concentrations useful for stock preparations.
  • Reported in vivo formulation options that yield clear solutions at ≥2.5 mg/mL.
  • Storage recommendations provided to maintain stability during short- and long-term storage.

Catalog No. 50-002-21019 Supplier Medchemexpress LLC Supplier No. HY145705MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triphen diol is a phenol diol small-molecule research compound (CAS 1213777-80-0) reported to induce apoptosis and inhibit proliferation in pancreatic cancer models. It has molecular formula C22H20O4, molecular weight 348.39 g/mol, and is supplied with high reported purity for use in in vitro and in vivo studies. Solubility and storage guidance is provided to support experimental workflows.

  • Phenol diol small-molecule with reported anticancer activity.
  • Demonstrated apoptosis induction and antiproliferative effects in pancreatic cancer cell lines.
  • High reported purity suitable for biological assays.
  • Soluble in DMSO at concentrations useful for stock preparations.
  • Reported in vivo formulation options that yield clear solutions at ≥2.5 mg/mL.
  • Storage recommendations provided to maintain stability during short- and long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.