Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2H-1-benzopyran-7-ol, 3,4-dihydro-3-(4-hydroxyphenyl)-4-(4-methoxyphenyl)-, (3R,4S)-rel- | 1213777-80-0 | 99.9% | C22H20O4 | 10MG
SDP

Catalog No. 5000259429
Encompass_Preferred

Triphen diol is a phenol diol derivative reported to have anticancer activity against pancreatic cancer and cholangiocarcinoma. It induces apoptosis through caspase-mediated and caspase-independent mechanisms and is supplied as a high-purity research reagent for laboratory studies.

  • Phenol diol derivative with reported anticancer activity.
  • Induces apoptosis via caspase-mediated and caspase-independent pathways.
  • High purity (~99.9%) suitable for research applications.
  • Molecular formula C22H20O4; molecular weight 348.39 g·mol⁻¹.
  • Soluble in DMSO (≈100 mg/mL); use freshly opened solvent to ensure solubility.
  • Store protected from light; in solvent store short-term at -20°C and long-term at -80°C.

Catalog No. 50-002-59429 Supplier Medchemexpress LLC Supplier No. HY1457010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Triphen diol is a phenol diol derivative reported to have anticancer activity against pancreatic cancer and cholangiocarcinoma. It induces apoptosis through caspase-mediated and caspase-independent mechanisms and is supplied as a high-purity research reagent for laboratory studies.

  • Phenol diol derivative with reported anticancer activity.
  • Induces apoptosis via caspase-mediated and caspase-independent pathways.
  • High purity (~99.9%) suitable for research applications.
  • Molecular formula C22H20O4; molecular weight 348.39 g·mol⁻¹.
  • Soluble in DMSO (≈100 mg/mL); use freshly opened solvent to ensure solubility.
  • Store protected from light; in solvent store short-term at -20°C and long-term at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.