Learn More
Medchemexpress LLC 2H-1-benzopyran-7-ol derivative (2R enantiomer) | 1443752-77-9 | 98.2% | 457.56 | C29H31NO4 | 5MG
SDP
Catalog No. 5000238328
Encompass_Preferred
(R)-OP-1074 is the R-enantiomer of OP-1074, a research-grade selective estrogen receptor degrader (PA-SERD) with potent anti-estrogenic activity against ERα and ERβ. It is supplied as a solid for research use and can be used as a control compound in estrogen receptor signaling studies. CAS 1443752-77-9; formula C29H31NO4; molecular weight 457.56.
Catalog No. 50-002-38328 Supplier Medchemexpress LLC Supplier No. HY125263B5MG
(R)-OP-1074 is the R-enantiomer of OP-1074, a research-grade selective estrogen receptor degrader (PA-SERD) with potent anti-estrogenic activity against ERα and ERβ. It is supplied as a solid for research use and can be used as a control compound in estrogen receptor signaling studies. CAS 1443752-77-9; formula C29H31NO4; molecular weight 457.56.