Learn More
Cayman Chemical 2H,2H,3H,3H-Perfluorodecanoic Acid, Purity: ≥80%, CAS: 812-70-4, 50 mg
SDP
Catalog No. 502917575
Encompass_Preferred
A polyfluoroalkyl substance; has been found as a contaminant in freshwater reserves and marine seafood
Catalog No. 50-291-7575 Supplier Cayman Chemical Supplier No. 3727150 MG
A polyfluoroalkyl substance; has been found as a contaminant in freshwater reserves and marine seafood