Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2H,2H,3H,3H-perfluorooctanoic acid | 914637-49-3 | MFCD08460490 | 99.8% | 342.11 g/mol | C8H5F11O2 | 50 MG
SDP

Catalog No. 5000208391
Encompass_Preferred

2H,2H,3H,3H-perfluorooctanoic acid is a fluorinated carboxylic acid used as an analytical reference standard and intermediate in environmental and fluorochemical research.

  • High purity 99.8% (GC)
  • Cas number 914637-49-3
  • Molecular formula C8H5F11O2
  • Molecular weight 342.11 g/mol
  • Suitable as an analytical reference standard for PFAS and environmental testing
  • Available in 50 MG quantity with accompanying COA

Catalog No. 50-002-08391 Supplier Medchemexpress LLC Supplier No. HYW08108550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2H,2H,3H,3H-perfluorooctanoic acid is a fluorinated carboxylic acid used as an analytical reference standard and intermediate in environmental and fluorochemical research.

  • High purity 99.8% (GC)
  • Cas number 914637-49-3
  • Molecular formula C8H5F11O2
  • Molecular weight 342.11 g/mol
  • Suitable as an analytical reference standard for PFAS and environmental testing
  • Available in 50 MG quantity with accompanying COA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.