Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2H,3H-Perfluoropentane, tech. 90%, Thermo Scientific Chemicals

Catalog No. AAB2163306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2163306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2163306 Supplier Thermo Scientific Chemicals Supplier No. B2163306

Used as a zero ozone-depletion alternative to hydro chlorofluorocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a zero ozone-depletion alternative to hydro chlorofluorocarbons.

Solubility
Soluble in water (140 mg/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138495-42-8
Molecular Formula C5H2F10
Molecular Weight (g/mol) 252.06
MDL Number MFCD00798129
InChI Key RIQRGMUSBYGDBL-UHFFFAOYNA-N
Synonym 2h,3h-decafluoropentane, vertrel xf, 2h,3h-perfluoropentane, hfc-4310mee, 1,1,1,2,3,4,4,5,5,5-decafluoropentane, 2,3-dihydroperfluoropentane, pentane, 1,1,1,2,2,3,4,5,5,5-decafluoro, hfc-43-10mee, 2,3-dihydrodecafluoropentane, hfc-43
PubChem CID 86240
IUPAC Name 1,1,1,2,2,3,4,5,5,5-decafluoropentane
SMILES FC(C(F)C(F)(F)C(F)(F)F)C(F)(F)F

Specifications

Melting Point -80°C
Density 1.6
Boiling Point 54°C
Flash Point None
Refractive Index <1.3000
Linear Formula CF3CHFCHFCF2CF3
Quantity 5 g
Solubility Information Soluble in water (140mg/L at 25°C).
Formula Weight 252.05
Percent Purity 90%
Grade Technical
Chemical Name or Material 2H,3H-Perfluoropentane
Show More Show Less
Hazard Category H315-H319-H335-H373
Precautionary Statement P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.