Learn More
Medchemexpress LLC 2H-benzimidazole-2-thione, 5-chloro-1,3-dihydro- | 25369-78-2 | MFCD01658762 | 99.8% | 184.65 g/mol | C7H5ClN2S | 25 G
SDP
Catalog No. 5000304023
Encompass_Preferred
5-Chloro-2-mercaptobenzimidazole is a research-grade tyrosinase inhibitor used to study melanogenesis, oxidative stress, and related biochemical pathways. It exhibits low-nanomolar inhibitory activity against tyrosinase substrates and shows antioxidant and anti-melanogenic effects, supplied as a high-purity solid for in vitro and in vivo research.
  • Selective tyrosinase inhibitor with low-nanomolar IC50s against L-tyrosine and L-dopa.
  • Demonstrates antioxidant and anti-melanogenic activity in cellular assays.
  • High purity (~99.8%) suitable for biochemical and cell-based research.
  • Soluble in DMSO; manufacturer provides protocols for in vivo formulations.
  • Stable as a solid under recommended cold storage conditions.
Catalog No. 50-003-04023 Supplier Medchemexpress LLC Supplier No. 25GHYW037976
5-Chloro-2-mercaptobenzimidazole is a research-grade tyrosinase inhibitor used to study melanogenesis, oxidative stress, and related biochemical pathways. It exhibits low-nanomolar inhibitory activity against tyrosinase substrates and shows antioxidant and anti-melanogenic effects, supplied as a high-purity solid for in vitro and in vivo research.
  • Selective tyrosinase inhibitor with low-nanomolar IC50s against L-tyrosine and L-dopa.
  • Demonstrates antioxidant and anti-melanogenic activity in cellular assays.
  • High purity (~99.8%) suitable for biochemical and cell-based research.
  • Soluble in DMSO; manufacturer provides protocols for in vivo formulations.
  • Stable as a solid under recommended cold storage conditions.