Learn More
Medchemexpress LLC 2H-Cyclopenta[b]furan-2-one, 5-(benzoyloxy)hexahydro-4-(hydroxymethyl)-, (3aR,4S,5R,6aS)- | 39746-00-4 | 98.0% | 276.28 | 10 G
SDP
Catalog No. 5000404186
Encompass_Preferred
(-)-Corey lactone benzoate is a building block in the chemical synthesis of corey aldehyde. It is for research use only and not sold to patients. It appears as a solid with a white to off-white color.
  • Building block in the chemical synthesis of corey aldehyde.
  • For research use only.
  • Appears as a solid.
  • White to off-white color.
Catalog No. 50-004-04186 Supplier Medchemexpress LLC Supplier No. HY2000310G
(-)-Corey lactone benzoate is a building block in the chemical synthesis of corey aldehyde. It is for research use only and not sold to patients. It appears as a solid with a white to off-white color.
  • Building block in the chemical synthesis of corey aldehyde.
  • For research use only.
  • Appears as a solid.
  • White to off-white color.