Learn More
Medchemexpress LLC 2H-Furo[2,3-h]-1-benzopyran-2-one | 523-50-2 | 186.16 | 50 MG
SDP
Catalog No. 5000415843
Encompass_Preferred
Angelicin is a furanocoumarin compound that inhibits NF-κB and MAPK signaling pathways, exhibiting anti-inflammatory, antiviral, and antitumor activities. It suppresses the lytic replication of γ-herpesviruses and induces apoptosis in neuroblastoma cells by downregulating anti-apoptotic proteins and activating caspases.
  • Inhibits NF-κB and MAPK signaling pathways.
  • Exhibits anti-inflammatory, antiviral, and antitumor activities.
  • Suppresses lytic replication of γ-herpesviruses.
  • Induces apoptosis in neuroblastoma cells.
  • Downregulates anti-apoptotic proteins like Bcl-2, Bcl-xL, and Mcl-1.
  • Activates caspase-9 and caspase-3.
Catalog No. 50-004-15843 Supplier Medchemexpress LLC Supplier No. HYN076350MG
Angelicin is a furanocoumarin compound that inhibits NF-κB and MAPK signaling pathways, exhibiting anti-inflammatory, antiviral, and antitumor activities. It suppresses the lytic replication of γ-herpesviruses and induces apoptosis in neuroblastoma cells by downregulating anti-apoptotic proteins and activating caspases.
  • Inhibits NF-κB and MAPK signaling pathways.
  • Exhibits anti-inflammatory, antiviral, and antitumor activities.
  • Suppresses lytic replication of γ-herpesviruses.
  • Induces apoptosis in neuroblastoma cells.
  • Downregulates anti-apoptotic proteins like Bcl-2, Bcl-xL, and Mcl-1.
  • Activates caspase-9 and caspase-3.