Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2H-Indazole-7-carboxamide, 2-[4-[1-[17-[4-[[(4R,5S)-4,5-bis(4-chlorophenyl)-4,5-dihydro-2-[ | 00-00-0 | MFCD32705102 | 99.0% | 1145.09 | C58H63Cl2N11O10 | 5 MG
SDP

Catalog No. 5000264035
Encompass_Preferred

rel-PROTAC PARP1 degrader is a research-grade PROTAC small molecule that induces proteasomal degradation of PARP1 via an MDM2 E3 ligase ligand. It is intended for in vitro and cellular studies of PARP1 biology and targeted protein degradation.

  • Induces PARP1 degradation via MDM2 E3 ligase
  • Purity 98.99%
  • Molecular weight 1145.09
  • Molecular formula C58H63Cl2N11O10
  • CAS number 2369022-68-2
  • Available in research pack sizes including 1 mg, 5 mg, and 10 mg
  • Intended for research use only

Catalog No. 50-002-64035 Supplier Medchemexpress LLC Supplier No. HY114324A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

rel-PROTAC PARP1 degrader is a research-grade PROTAC small molecule that induces proteasomal degradation of PARP1 via an MDM2 E3 ligase ligand. It is intended for in vitro and cellular studies of PARP1 biology and targeted protein degradation.

  • Induces PARP1 degradation via MDM2 E3 ligase
  • Purity 98.99%
  • Molecular weight 1145.09
  • Molecular formula C58H63Cl2N11O10
  • CAS number 2369022-68-2
  • Available in research pack sizes including 1 mg, 5 mg, and 10 mg
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.