Learn More
Medchemexpress LLC 2H-indol-2-one, 1,3-dihydro-3-(2-pyridinylmethylene)-(3Z)- | 187325-53-7 | 98.0% | 222.25 g/mol | C14H10N2O | 50MG
SDP
Catalog No. 5000234922
Encompass_Preferred
GSK-3β inhibitor 1 is a small-molecule inhibitor of glycogen synthase kinase-3 beta (CAS 187325-53-7) with low-nanomolar biochemical potency and reported antidiabetic efficacy in obese streptozotocin-treated rats. The compound is provided as a solid for research use and is soluble in DMSO with recommended storage conditions for powder and solutions.
Catalog No. 50-002-34922 Supplier Medchemexpress LLC Supplier No. HY12614450MG
GSK-3β inhibitor 1 is a small-molecule inhibitor of glycogen synthase kinase-3 beta (CAS 187325-53-7) with low-nanomolar biochemical potency and reported antidiabetic efficacy in obese streptozotocin-treated rats. The compound is provided as a solid for research use and is soluble in DMSO with recommended storage conditions for powder and solutions.