Learn More
Medchemexpress LLC 2H-Indol-2-one, 1,3-dihydro-3-hydroxy-3-(2-oxo-2-phenylethyl)-1-(2-phenylethyl)- | 421578-93-0 | 99.8% | C24H21NO3 | 25 MG
SDP
Catalog No. 5000352708
Encompass_Preferred
GLP-1R modulator C5 is an allosteric modulator enhancing GLP-1 binding to GLP-1R via a transmembrane site (EC50 1.59 ± 0.53 μM).
  • Enhances GLP-1 binding to GLP-1R.
  • Acts via a transmembrane site.
  • High purity of 99.8%.
  • White to off-white solid.
  • Suitable for research applications.
Catalog No. 50-003-52708 Supplier Medchemexpress LLC Supplier No. 25MGHY141840
GLP-1R modulator C5 is an allosteric modulator enhancing GLP-1 binding to GLP-1R via a transmembrane site (EC50 1.59 ± 0.53 μM).
  • Enhances GLP-1 binding to GLP-1R.
  • Acts via a transmembrane site.
  • High purity of 99.8%.
  • White to off-white solid.
  • Suitable for research applications.