Learn More
Medchemexpress LLC 2H-Indol-2-one, 5-chloro-3-[(3,5-dimethyl-1H-pyrrol-2-yl)methylene]-1,3-dihydro-, (3Z)- | 1055412-47-9 | 95.7% | 272.73 | 25 MG
SDP
Catalog No. 5000384995
Encompass_Preferred
(Z)-SU5614 is a potent FLT3 inhibitor that selectively induces growth arrest, apoptosis, and cell cycle arrest in Ba/F3 and AML cell lines expressing a constitutively activated FLT3.
  • Potent FLT3 inhibitor.
  • Selectively induces growth arrest in Ba/F3 and AML cell lines expressing a constitutively activated FLT3.
  • Induces apoptosis in Ba/F3 and AML cell lines expressing a constitutively activated FLT3.
  • Induces cell cycle arrest in Ba/F3 and AML cell lines expressing a constitutively activated FLT3.
Catalog No. 50-003-84995 Supplier Medchemexpress LLC Supplier No. 25MGHY18952A
Add to Cart
Add to Cart
(Z)-SU5614 is a potent FLT3 inhibitor that selectively induces growth arrest, apoptosis, and cell cycle arrest in Ba/F3 and AML cell lines expressing a constitutively activated FLT3.
  • Potent FLT3 inhibitor.
  • Selectively induces growth arrest in Ba/F3 and AML cell lines expressing a constitutively activated FLT3.
  • Induces apoptosis in Ba/F3 and AML cell lines expressing a constitutively activated FLT3.
  • Induces cell cycle arrest in Ba/F3 and AML cell lines expressing a constitutively activated FLT3.