Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2H-Perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane, Thermo Scientific™

Catalog No. AAL1695306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1695306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AAL1695306 Supplier Thermo Scientific Chemicals Supplier No. L1695306
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 26738-51-2
Melting Point -115°C
Density 1.723
Boiling Point 192°C to 195°C
Flash Point None
Molecular Formula C14HF29O4
Quantity 5 g
MDL Number MFCD00054717
Synonym 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-icosafluoro-5,8,11-tris trifluoromethyl-3,6,9,12-tetraoxapentadecane, 3,6,9,12-tetraoxapentadecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-eicosafluoro-5,8,11-tris trifluoromethyl, freon e 4, freon e-4, 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane, 1,1,1,2,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy-3-1,2,2,2-tetrafluoroethoxy propane
InChI Key NOCLWIPRQIWFMR-UHFFFAOYNA-N
SMILES FC(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F
IUPAC Name 1,1,1,2,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]-3-(1,2,2,2-tetrafluoroethoxy)propane
Molecular Weight (g/mol) 784.11
PubChem CID 117327
Formula Weight 784.12
Chemical Name or Material 2H-Perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane
Show More Show Less
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.