Learn More
Medchemexpress LLC 2H-pyran-2,4(3H)-dione, 3-acetyl-6-methyl- | 520-45-6 | MFCD00066709 | 99.9% | 1 G
SDP
Catalog No. 5000413621
Encompass_Preferred
Dehydroacetic acid (Standard) is the analytical standard of Dehydroacetic acid. This pyrone derivative is intended for research and analytical applications, exhibiting antibacterial, antifungal, and phytotoxic activities. It finds use in the preservation of cosmetics, food, and beverages, and serves as a crucial component in synthesizing various heterocycles and cycloaddition products. As a reference standard, it is commonly applied in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Pyrone derivative.
  • Exhibits antibacterial, antifungal, and phytotoxic activities.
  • Suitable for preservation in cosmetics, food, and beverages.
  • Used in the synthesis of various heterocycles and cycloaddition products.
  • Reference standard for qualitative, quantitative, and methodological research (HPLC, GC, MS).
Catalog No. 50-004-13621 Supplier Medchemexpress LLC Supplier No. HYB12111G
Add to Cart
Add to Cart
Dehydroacetic acid (Standard) is the analytical standard of Dehydroacetic acid. This pyrone derivative is intended for research and analytical applications, exhibiting antibacterial, antifungal, and phytotoxic activities. It finds use in the preservation of cosmetics, food, and beverages, and serves as a crucial component in synthesizing various heterocycles and cycloaddition products. As a reference standard, it is commonly applied in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Pyrone derivative.
  • Exhibits antibacterial, antifungal, and phytotoxic activities.
  • Suitable for preservation in cosmetics, food, and beverages.
  • Used in the synthesis of various heterocycles and cycloaddition products.
  • Reference standard for qualitative, quantitative, and methodological research (HPLC, GC, MS).