Learn More
Medchemexpress LLC 2H-Pyrido[1,2-a]pyrazine-1,6-dione, 3,4-dihydro-3-methyl-7-(4-methyl-1H-imidazol-1-yl)- | 1493797-28-6 | 99.7% | 469.46 | 1 ML
SDP
Catalog No. 5000386525
Encompass_Preferred
GSM III is a γ-Secretase complexe (GSEC) modulator. It significantly regulates amyloid-β (Aβ) length at the extracellular interface between the protease (NCT, PSEN) and the substrate APPC99. This product can be used for Alzheimer's disease (AD) research. It is for research use only.
  • Acts as a γ-Secretase complexe (GSEC) modulator
  • Regulates amyloid-β (Aβ) length
  • Is suitable for Alzheimer's disease (AD) research
Catalog No. 50-003-86525 Supplier Medchemexpress LLC Supplier No. HY17144410MM/1ML
GSM III is a γ-Secretase complexe (GSEC) modulator. It significantly regulates amyloid-β (Aβ) length at the extracellular interface between the protease (NCT, PSEN) and the substrate APPC99. This product can be used for Alzheimer's disease (AD) research. It is for research use only.
  • Acts as a γ-Secretase complexe (GSEC) modulator
  • Regulates amyloid-β (Aβ) length
  • Is suitable for Alzheimer's disease (AD) research