Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2H-Pyrido[3,4-b]indole-2,3-dicarboxylic acid, 1,3,4,9-tetrahydro-, 2-(9H-fluoren-9-ylmethyl) ester, (3S)- | 204322-23-6 | 99.2% | 438.47 | 25 G
SDP

Catalog No. 5000348260
Encompass_Preferred

Fmoc-Tpi-OH is a biologically active amino acid that can be used to synthesize antimicrobial peptide mimetics. This product appears as a white to off-white solid with a purity of 99.24% by HPLC. It is soluble in DMSO at 66.67 mg/mL.

  • Biologically active amino acid
  • Suitable for synthesizing antimicrobial peptide mimetics
  • White to off-white solid appearance
  • Purity of 99.24%
  • Soluble in DMSO (66.67 mg/mL)
  • Store at 4°C, protected from light

Catalog No. 50-003-48260 Supplier Medchemexpress LLC Supplier No. 25GHY138546
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Tpi-OH is a biologically active amino acid that can be used to synthesize antimicrobial peptide mimetics. This product appears as a white to off-white solid with a purity of 99.24% by HPLC. It is soluble in DMSO at 66.67 mg/mL.

  • Biologically active amino acid
  • Suitable for synthesizing antimicrobial peptide mimetics
  • White to off-white solid appearance
  • Purity of 99.24%
  • Soluble in DMSO (66.67 mg/mL)
  • Store at 4°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.