Learn More
Medchemexpress LLC 2H-tetrazole-2-butanoic acid, β-amino-5-[4-[(5-chloro-3-fluoro-2-pyridinyl)oxy]phenyl]-, (βS)- | 1799681-85-8 | 98.4% | C16H14ClFN6O3 | 10MG
SDP
Catalog No. 5000261361
Encompass_Preferred
LYS006 is a small-molecule inhibitor of leukotriene A4 hydrolase (LTA4H) developed for research into inflammatory and autoimmune disorders. It is reported to have subnanomolar to low-nanomolar potency (IC50 ≈ 2 nM) and is supplied with analytical characterization suitable for laboratory studies.
  • Potent leukotriene A4 hydrolase (LTA4H) inhibitor (IC50 ≈ 2 nM).
  • High purity suitable for research (≈98.4%).
  • Available as solid and as DMSO solution for assay flexibility.
  • Characterized by a certificate of analysis with structural confirmation.
  • Intended for preclinical and pharmacology studies in inflammation and immunology.
Catalog No. 50-002-61361 Supplier Medchemexpress LLC Supplier No. HY13729810MG
LYS006 is a small-molecule inhibitor of leukotriene A4 hydrolase (LTA4H) developed for research into inflammatory and autoimmune disorders. It is reported to have subnanomolar to low-nanomolar potency (IC50 ≈ 2 nM) and is supplied with analytical characterization suitable for laboratory studies.
  • Potent leukotriene A4 hydrolase (LTA4H) inhibitor (IC50 ≈ 2 nM).
  • High purity suitable for research (≈98.4%).
  • Available as solid and as DMSO solution for assay flexibility.
  • Characterized by a certificate of analysis with structural confirmation.
  • Intended for preclinical and pharmacology studies in inflammation and immunology.