Learn More
Cayman Chemical 2HChoArgtrIfluoroacetAT 10mg
SDP
Catalog No. 502936271
Encompass_Preferred
A steroid-based cationic lipid that can be used to facilitate gene transfection; forms a complex with pDNA and decreases pDNA migration in an electrophoretic mobility shift assay at +/- charge ratios of 4 or higher; facilitates transfection of the luciferase gene into H1299 cells, which can be inhibited by methyl-β-cyclodextrin and genistein; induces cytotoxicity in H1299 cells (IC50 = 92.7 μg/ml)
Catalog No. 50-293-6271 Supplier Cayman Chemical Supplier No. 2594410 MG
A steroid-based cationic lipid that can be used to facilitate gene transfection; forms a complex with pDNA and decreases pDNA migration in an electrophoretic mobility shift assay at +/- charge ratios of 4 or higher; facilitates transfection of the luciferase gene into H1299 cells, which can be inhibited by methyl-β-cyclodextrin and genistein; induces cytotoxicity in H1299 cells (IC50 = 92.7 μg/ml)