Learn More
Perkin Elmer US LLC 2-Hole Graphite Short Ferrules, 0.0625 in x 0.5 mm I.D., Pkg. 10
Catalog No. 502111833
Encompass_Preferred
Of the three ferrule types available, graphite is the softest, making it less damaging to capillary columns when overtightened. Graphite seals easily and does not stick to glass columns. These one-piece ferrules do not require rear ferrules. Quantity: Package of 10 Nut Size: 1/16 inch Column Inner Diameter: 0.32 mm Ferrule Inner Diameter: 0.5 mm
Catalog No. 50-211-1833 Supplier Perkin Elmer US LLC Supplier No. 09200788
Of the three ferrule types available, graphite is the softest, making it less damaging to capillary columns when overtightened. Graphite seals easily and does not stick to glass columns. These one-piece ferrules do not require rear ferrules. Quantity: Package of 10 Nut Size: 1/16 inch Column Inner Diameter: 0.32 mm Ferrule Inner Diameter: 0.5 mm