Learn More
Cayman Chemical 2-hydroxy Docosanoic Acid methyl ester, Purity: ≥98%, CAS: 13980-17-1, 25 mg
SDP
Catalog No. 502936340
Encompass_Preferred
A hydroxy fatty acid methyl ester; has been found in ripe and unripe strawberry homogenates, Pseudosuberites and S. massa sea sponges, sediment samples from the Harney River, and C. frondosa and D. cinerea extracts
Catalog No. 50-293-6340 Supplier Cayman Chemical Supplier No. 2459625 MG
A hydroxy fatty acid methyl ester; has been found in ripe and unripe strawberry homogenates, Pseudosuberites and S. massa sea sponges, sediment samples from the Harney River, and C. frondosa and D. cinerea extracts