Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Hydroxymethyl-18-Crown-6

Catalog No. 388440250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-844-0250MG 250 mg
38-844-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 38-844-0250MG Supplier Sigma Aldrich Supplier No. 388440250MG
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Hydroxymethyl-18-Crown-6

Specifications

CAS 70069-04-4
Density 1.174 g/mL at 25°C
Boiling Point 150°C (0.5 mmHg, lit.)
Molecular Formula C13H26O7
Refractive Index n20/D 1.479
Linear Formula C13H26O7
MDL Number MFCD00188046
Quantity 250 mg
Synonym 1,4,7,10,13,16-Hexaoxacyclooctadecane-2-methanol; 18-Crown-6-methanol
Molecular Weight (g/mol) 294.34 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.