Learn More
Cayman Chemical 2-hydroxy Palmitic Acid methyl ester, Purity: ≥98%, CAS: 16742-51-1, 50 mg
SDP
Catalog No. 502935614
Encompass_Preferred
A hydroxy fatty acid methyl ester; a minor component in essential oils of P. sylvestris and B. javanica extracts
Catalog No. 50-293-5614 Supplier Cayman Chemical Supplier No. 2458950 MG
A hydroxy fatty acid methyl ester; a minor component in essential oils of P. sylvestris and B. javanica extracts