Learn More
Cayman Chemical 2-Imino-4-methylpiperidine (acetate), Purity: ≥98%, CAS: 165383-72-2, 50 mg
SDP
Catalog No. 502894094
Encompass_Preferred
2-Imino-4-methylpiperidine is a potent, general NOS inhibitor. It exhibits IC50 values of 0.1, 1.1, and 0.2 µM for inhibition of human iNOS, eNOS, and nNOS, respectively, at an arginine concentration of 30 µM.{5579}
Catalog No. 50-289-4094 Supplier Cayman Chemical Supplier No. 8101550 MG
2-Imino-4-methylpiperidine is a potent, general NOS inhibitor. It exhibits IC50 values of 0.1, 1.1, and 0.2 µM for inhibition of human iNOS, eNOS, and nNOS, respectively, at an arginine concentration of 30 µM.{5579}