Learn More
Hach Company 2in. CPVC Union Tee 1.5in pH
Catalog No. 5000207059 Shop All Hach Company Products
Encompass
2in CPVC Union Tee-for 1 5in pH/ORP SensorsCPVC 2-Inch Special Tee and Aluminum Junction Box for Use with LCP pH/ORP Sensors
Catalog No. 50-002-07059 Supplier Hach Company Supplier No. MH538N9A
2in CPVC Union Tee-for 1 5in pH/ORP SensorsCPVC 2-Inch Special Tee and Aluminum Junction Box for Use with LCP pH/ORP Sensors