Learn More
Ace Glass, Inc. 2in NPT PIPE NIPPLE
SDP
Catalog No. 5000854812
Encompass
On/Off ball valve with lockable handle (padlock not included). Wetted surfaces are stainless steel and PTFE.
Catalog No. 50-008-54812 Supplier Ace Glass, Inc. Supplier No. 1382410
On/Off ball valve with lockable handle (padlock not included). Wetted surfaces are stainless steel and PTFE.