Learn More
Hach Company 2in. PVC Union Tee 1.5in pH
Catalog No. 5000207080 Shop All Hach Company Products
Encompass
2in PVC Union Mounting Tee-For 1 5 inch pH/ORP sensorsPVC 2-Inch Special Tee and Aluminum Junction Box for Use with LCP pH/ORP Sensors
Catalog No. 50-002-07080 Supplier Hach Company Supplier No. MH578N9A
2in PVC Union Mounting Tee-For 1 5 inch pH/ORP sensorsPVC 2-Inch Special Tee and Aluminum Junction Box for Use with LCP pH/ORP Sensors