Learn More
Hach Company 2in. PVDF Union T 3700 w/o Box
Catalog No. 5000207110 Shop All Hach Company Products
Encompass
2in PVDF Union Tee-for 3700 Series Conductivity Sensors-without Junction BoxPVDF union adapter and standard 2-inch pipe tee for use with 3700-series inductive/electrodeless conductivity sensors
Catalog No. 50-002-07110 Supplier Hach Company Supplier No. MH568N3NZ
2in PVDF Union Tee-for 3700 Series Conductivity Sensors-without Junction BoxPVDF union adapter and standard 2-inch pipe tee for use with 3700-series inductive/electrodeless conductivity sensors