Learn More
Ace Glass, Inc. Flange clamp, 60mm(2in), SS, quick release
SDP
Catalog No. NC9934527
Encompass
60mm (2) stainless steel quick release flange clamp without support rod, for use with two-piece pressure reaction flasks and for heads with Schott flanges.
Catalog No. NC9934527 Supplier Ace Glass, Inc. Supplier No. 651722
60mm (2) stainless steel quick release flange clamp without support rod, for use with two-piece pressure reaction flasks and for heads with Schott flanges.