Learn More
Accuflow Systems Steel & O'Brien Tri-Clamp Adapter, 2 in, 3/8 in Barb
Catalog No. NC3666349
Encompass_Preferred
Tri-clamp to hose barb adapter designed to connect 2 in tri-clamp process lines to 3/8 in ID rubber hose. The fitting provides a sanitary clamp connection with a grooved barb for secure hose retention and hygienic fluid transfer in food, beverage, brewery, dairy, and laboratory process lines.
  • Connects 2 in tri-clamp to 3/8 in hose barb.
  • Constructed from stainless steel; commonly offered in 304 or 316L.
  • Sanitary design for hygienic fluid transfer.
  • Grooved hose barb for secure hose retention.
  • Suitable for food, beverage, dairy, brewery, and biotech process lines.
  • Available in multiple clamp and barb size combinations.
Catalog No. NC3666349 Supplier Accuflow Systems Supplier No. 14MPHR2X375S
Tri-clamp to hose barb adapter designed to connect 2 in tri-clamp process lines to 3/8 in ID rubber hose. The fitting provides a sanitary clamp connection with a grooved barb for secure hose retention and hygienic fluid transfer in food, beverage, brewery, dairy, and laboratory process lines.
  • Connects 2 in tri-clamp to 3/8 in hose barb.
  • Constructed from stainless steel; commonly offered in 304 or 316L.
  • Sanitary design for hygienic fluid transfer.
  • Grooved hose barb for secure hose retention.
  • Suitable for food, beverage, dairy, brewery, and biotech process lines.
  • Available in multiple clamp and barb size combinations.