Learn More
McMaster-Carr High-Density Thread Sealant Tape, PTFE, 0.0032" Thick, 2" Wide, 14 Yard Long, White
Catalog No. NC1442066
Encompass
This tape has a higher density than standard tape for a better seal between threads. Made of PTFE, it has a slippery surface that keeps threads from binding to make parts easy to assemble.
Catalog No. NC1442066 Supplier McMaster-Carr Supplier No. 6802K77
This tape has a higher density than standard tape for a better seal between threads. Made of PTFE, it has a slippery surface that keeps threads from binding to make parts easy to assemble.