Learn More
Cayman Chemical 2-(Isopentylamino)naphthalene-1,4-dione, Purity: ≥98%, CAS: 1607447-79-9, 25 mg
SDP
Catalog No. 502990133
Encompass_Preferred
A vitamin K analog; inhibits PTZ-induced spasms and MES-induced tonic hindlimb extension in mice (ED50s = 349.2 and 108.1 mg/kg, respectively); protects mice against seizures in the 6 Hz psychomotor seizure test (ED50s = 152.7 and 263.7 mg/kg at stimulus intensities of 32 and 44 mA, respectively)
Catalog No. 50-299-0133 Supplier Cayman Chemical Supplier No. 3110725 MG
A vitamin K analog; inhibits PTZ-induced spasms and MES-induced tonic hindlimb extension in mice (ED50s = 349.2 and 108.1 mg/kg, respectively); protects mice against seizures in the 6 Hz psychomotor seizure test (ED50s = 152.7 and 263.7 mg/kg at stimulus intensities of 32 and 44 mA, respectively)