Learn More
Ace Glass, Inc. 2 liter epoxy coated flash chromatography reservoir with35/20 upper joint and 35/20 lower joint.
SDP
Catalog No. 50978570
Encompass_Preferred
2L epoxy coated flash chromatography reservoir with 35/20 upper joint and lower joints. Reservoir connects between the column and flow control adapter.
Catalog No. 50-978-570 Supplier Ace Glass, Inc. Supplier No. 587245
2L epoxy coated flash chromatography reservoir with 35/20 upper joint and lower joints. Reservoir connects between the column and flow control adapter.