Learn More
Ace Glass, Inc. Beaker, 2L, jacketed, Ace-Safe hose connections for 3/8intubing, 190mm inside height, 119mm inside diameter
SDP
Catalog No. 50971851
Encompass_Preferred
2L jacketed beaker with one upper and one lower Ace-Safe hose connection on opposite sides, for 3/8 in. tubing. Dimensions: 190mm I.H. x 119mm I.D. Does not have a pouring spout.
Catalog No. 50-971-851 Supplier Ace Glass, Inc. Supplier No. 5340120
Add to Cart
Add to Cart
2L jacketed beaker with one upper and one lower Ace-Safe hose connection on opposite sides, for 3/8 in. tubing. Dimensions: 190mm I.H. x 119mm I.D. Does not have a pouring spout.