Learn More
CELLTREAT Scientific Products 2L Erlenmeyer Flask, Vent Cap, Baffled Bottom, Sterile
SDP
Catalog No. 50202096
Encompass_Preferred
CELLTREAT Scientific Products 2L Erlenmeyer Flask, Baffled Bottom. Designed for mammalian suspension culture applications as well as bacterial and yeast culture. Conveniently pre-sterilized. 70mm opening for easy filling and dispensing. Over-sized vent area in cap for improved gas exchange. Baffled bottom option improves mixing and aeration. Vent cap with baffled bottom and maximum volum of 2200mL. Non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 6/case.
Catalog No. 50-202-096 Supplier CELLTREAT Scientific Products Supplier No. 229855
CELLTREAT Scientific Products 2L Erlenmeyer Flask, Baffled Bottom. Designed for mammalian suspension culture applications as well as bacterial and yeast culture. Conveniently pre-sterilized. 70mm opening for easy filling and dispensing. Over-sized vent area in cap for improved gas exchange. Baffled bottom option improves mixing and aeration. Vent cap with baffled bottom and maximum volum of 2200mL. Non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 6/case.