Learn More
CELLTREAT Scientific Products 2L Fernbach Flask, Vent Cap, Baffled Bottom, Sterile
SDP
Catalog No. 50202098
Encompass_Preferred
CELLTREAT Scientific Products 2L Fernbach Flask, Baffled Bottom. Designed for mammalian suspension culture applications as well as bacterial and yeast culture. Conveniently pre-sterilized. 70mm opening for easy filling and dispensing. Over-sized vent area in cap for improved gas exchange. Fernbach flask shape increases surface area to volume ratio. Baffled bottom option improves mixing and aeration. Vent cap with baffled bottom and maximum volume of 2400mL. Non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 6/case.
Catalog No. 50-202-098 Supplier CELLTREAT Scientific Products Supplier No. 229865
CELLTREAT Scientific Products 2L Fernbach Flask, Baffled Bottom. Designed for mammalian suspension culture applications as well as bacterial and yeast culture. Conveniently pre-sterilized. 70mm opening for easy filling and dispensing. Over-sized vent area in cap for improved gas exchange. Fernbach flask shape increases surface area to volume ratio. Baffled bottom option improves mixing and aeration. Vent cap with baffled bottom and maximum volume of 2400mL. Non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 6/case.