Learn More
Ace Glass, Inc. 2L filter reactor body with 100mm ID grooved top flange, #80 bottom thread, 130mm IDx180mm depth, with CAPFE O-ring. Pressure rated at 35 psi.
SDP
Catalog No. 50972789
Encompass
Catalog No. 50-972-789 Supplier Ace Glass, Inc. Supplier No. 638419