Learn More
Bellco Glass, Inc. 2L Erlenmeyer Flask, Vent Cap, Baffled Bottom, Sterile
SDP
Catalog No. 502329627
Encompass_Preferred
Designed for mammalian suspension culture applications as well as bacterial and yeast cultureConveniently pre-sterilized70mm opening for easy filling and dispensingOver-sized vent area in cap for improved gas exchangeBaffled bottom option improves mixing and aeration Pack Description1/BagCase Quantity6SterileYes, bottleNon-pyrogenicYesAutoclavableYesPrimary ColorFlask: ClearSecondary ColorCap: GreenPrimary MaterialFlask: PolycarbonateSecondary MaterialCap: PolypropyleneMaximum Volume2200mLWorking Volume400mLGraduatedYesTissue Culture TreatedNo
Catalog No. 50-232-9627 Supplier Bellco Glass, Inc. Supplier No. 229855000
Designed for mammalian suspension culture applications as well as bacterial and yeast cultureConveniently pre-sterilized70mm opening for easy filling and dispensingOver-sized vent area in cap for improved gas exchangeBaffled bottom option improves mixing and aeration Pack Description1/BagCase Quantity6SterileYes, bottleNon-pyrogenicYesAutoclavableYesPrimary ColorFlask: ClearSecondary ColorCap: GreenPrimary MaterialFlask: PolycarbonateSecondary MaterialCap: PolypropyleneMaximum Volume2200mLWorking Volume400mLGraduatedYesTissue Culture TreatedNo