Learn More
CELLTREAT Scientific Products 2L Erlenmeyer Flask, Vent Cap, Plain Bottom, Sterile
SDP
Catalog No. 50202095
Encompass_Preferred
CELLTREAT Scientific Products 2L Erlenmeyer Flask. Designed for mammalian suspension culture applications as well as bacterial and yeast culture. Conveniently pre-sterilized. 70mm opening for easy filling and dispensing. Over-sized vent area in cap for improved gas exchange. Vent cap, plain bottom, with maximum volume of 2200mL. Non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 6/case.
Catalog No. 50-202-095 Supplier CELLTREAT Scientific Products Supplier No. 229850
CELLTREAT Scientific Products 2L Erlenmeyer Flask. Designed for mammalian suspension culture applications as well as bacterial and yeast culture. Conveniently pre-sterilized. 70mm opening for easy filling and dispensing. Over-sized vent area in cap for improved gas exchange. Vent cap, plain bottom, with maximum volume of 2200mL. Non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 6/case.