Learn More
Chemglass Life Sciences Chemglass 2 L Erlenmeyer Flask, High Efficiency, Polycarbonate, Sterile, Solid Cap
SDP
Catalog No. 502733467
Encompass
The Chemglass 2 L Erlenmeyer flask is a high-efficiency, medical-grade polycarbonate culture vessel designed for single-use bioprocessing and laboratory applications. It is sterile (E-beam), meets USP Class VI and ISO 10993/USP<661> compatibility standards, and is tested for tightness, drop resistance, endotoxins, and cell-culture suitability.
  • High transparency and clear graduation scale for easy observation.
  • Medical-grade polycarbonate construction with impact and oxidation resistance.
  • Sterile and non-pyrogenic; sterilized by E-beam.
  • Meets USP Class VI and ISO 10993/USP<661> compatibility requirements.
  • Withstands temperatures up to 121 °C.
  • Tested for tightness, drop resistance, endotoxin, DNA/RNA enzyme-free, and cell-culture suitability.
  • Packaged as sterile nests, four per case.
Catalog No. 50-273-3467 Supplier Chemglass Life Sciences Supplier No. CGN21652LSC
The Chemglass 2 L Erlenmeyer flask is a high-efficiency, medical-grade polycarbonate culture vessel designed for single-use bioprocessing and laboratory applications. It is sterile (E-beam), meets USP Class VI and ISO 10993/USP<661> compatibility standards, and is tested for tightness, drop resistance, endotoxins, and cell-culture suitability.
  • High transparency and clear graduation scale for easy observation.
  • Medical-grade polycarbonate construction with impact and oxidation resistance.
  • Sterile and non-pyrogenic; sterilized by E-beam.
  • Meets USP Class VI and ISO 10993/USP<661> compatibility requirements.
  • Withstands temperatures up to 121 °C.
  • Tested for tightness, drop resistance, endotoxin, DNA/RNA enzyme-free, and cell-culture suitability.
  • Packaged as sterile nests, four per case.