Learn More
Chemglass Life Sciences 2000mL Erlenmeyer Flask, 38-430 GPI Screw Thread
SDP
Catalog No. CG154306
Encompass_Preferred
2000mL Erlenmeyer Flask, 38-430 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.
Catalog No. CG154306 Supplier Chemglass Life Sciences Supplier No. CG154306
2000mL Erlenmeyer Flask, 38-430 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.