Learn More
Ace Glass, Inc. Extraction apparatus, liquid from ether, 2L, complete with6740-04 condenser, Por B funnel tube, outer chamber and6965-22 Erlenmeyer flask
SDP
Catalog No. 50978890
Encompass_Preferred
2L extraction apparatus designed for the extracting of liquids with ether. The dispersion of the ether is accomplished with the use of a 70-100micron porosity ACE fritted disc, which is attached to the base of the funnel tube.
Catalog No. 50-978-890 Supplier Ace Glass, Inc. Supplier No. 684050
2L extraction apparatus designed for the extracting of liquids with ether. The dispersion of the ether is accomplished with the use of a 70-100micron porosity ACE fritted disc, which is attached to the base of the funnel tube.