Learn More
Foxx Life Sciences Foxx Life Science EZclear Erlenmeyer Shaker Flasks
Catalog No. NC2587298
Encompass_Preferred
Foxx Life Science EZclear Erlenmeyer Shaker Flasks are molded from virgin, optically clear, non-leaching polycarbonate resin. Our flasks retain clarity after gamma sterilization, and comply with stringent FDA and USP requirements. Sterile flasks are additionally certified as non-pyrogenic and DNase/RNase-free, making them ideal for cell cultures, microbial cultures, plants, media preparation, storage, and all other applications. They also fit all standard shaking incubator clamps and can be placed on any platform.
Catalog No. NC2587298 Supplier Foxx Life Sciences Supplier No. 2485142OEM
Foxx Life Science EZclear Erlenmeyer Shaker Flasks are molded from virgin, optically clear, non-leaching polycarbonate resin. Our flasks retain clarity after gamma sterilization, and comply with stringent FDA and USP requirements. Sterile flasks are additionally certified as non-pyrogenic and DNase/RNase-free, making them ideal for cell cultures, microbial cultures, plants, media preparation, storage, and all other applications. They also fit all standard shaking incubator clamps and can be placed on any platform.