Learn More
Thomas Scientific 2L Fernbach Flask, Vent Cap, Baffled Bottom, Sterile
SDP
Catalog No. 501461533
Encompass
Designed for suspension culture applications Constructed from optically clear, autoclavable polycarbonate 70mm opening for easy filling and dispensing Over-sized vent area in cap for improved gas exchange Fernbach flask shape increases surface area to volume ratio NEW – Unique 2L Fernbach now available Available with plain or baffled bottom All Erlenmeyer and Fernbach flasks are individually bagged and supplied sterile (gamma irradiated) NEW – Non-treated Suspension Culture Flask now available in 850mL size with Plug Seal C
Catalog No. 50-146-1533 Supplier Thomas Scientific Supplier No. 1219Z49
Designed for suspension culture applications Constructed from optically clear, autoclavable polycarbonate 70mm opening for easy filling and dispensing Over-sized vent area in cap for improved gas exchange Fernbach flask shape increases surface area to volume ratio NEW – Unique 2L Fernbach now available Available with plain or baffled bottom All Erlenmeyer and Fernbach flasks are individually bagged and supplied sterile (gamma irradiated) NEW – Non-treated Suspension Culture Flask now available in 850mL size with Plug Seal C