Learn More
Chemglass Life Sciences 2, 000mL Glassblowers Heavy Wall RBF Blank
SDP
Catalog No. 501531800
Encompass_Preferred
2,000mL Glassblowers Heavy Wall Round Bottomed Flask Blank. Heavy Wall blanks are 30% heavier than Chemglass's standard CG-618 series. Blanks are used in the fabrication of single or multi-neck round bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. 50-153-1800 Supplier Chemglass Life Sciences Supplier No. CG61811HW
2,000mL Glassblowers Heavy Wall Round Bottomed Flask Blank. Heavy Wall blanks are 30% heavier than Chemglass's standard CG-618 series. Blanks are used in the fabrication of single or multi-neck round bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.