Learn More
Microsolv Technology Corporation Bottle, Mobile Phase Reservoir, Glass, Clear, 2L, Screw Top, GL45 Threads. 1 EA.
Catalog No. 502270180
Encompass_Preferred
One of our Items, this Clear, Borosilicate Glass Bottle has a 2L "Total Fill Capacity" and is perfect as an HPLC Mobile Phase Reservoir. The small opening, one liter Capacity and White Enamel Graduations on the outside of the Bottle help you determine the Solvent use and prevent running dry. This Bottle can be used with MicroSolv Filter Degassers , other Degassers or disposable Filter Degassers. A Solid Screw Cap without a liner and a polypropylene Drip Ring are included. This Bottle is often used with the Mobile Phase Delivery System and is Not "Safety Coated".
This item is not returnable. View return policy
Catalog No. 50-227-0180 Supplier Microsolv Technology Corporation Supplier No. 588072L
One of our Items, this Clear, Borosilicate Glass Bottle has a 2L "Total Fill Capacity" and is perfect as an HPLC Mobile Phase Reservoir. The small opening, one liter Capacity and White Enamel Graduations on the outside of the Bottle help you determine the Solvent use and prevent running dry. This Bottle can be used with MicroSolv Filter Degassers , other Degassers or disposable Filter Degassers. A Solid Screw Cap without a liner and a polypropylene Drip Ring are included. This Bottle is often used with the Mobile Phase Delivery System and is Not "Safety Coated".