Learn More
Microsolv Technology Corporation Bottles, Mobile Phase Reservoir, Glass, Clear, 2L, Screw Top, GL45 Threads. 5/CS.
Catalog No. 502270181
Encompass_Preferred
Some of our Bottles, these Clear, Borosilicate Glass Bottles have a 2L "Total Fill Capacity" and are perfect as HPLC Mobile Phase Reservoirs. The small openings, one liter Capacity and White Enamel Graduations on the outside of the Bottles help you determine the Solvent use and prevent running dry. This Bottle can be used with MicroSolv Filter Degassers , other Degassers or disposable Filter Degassers. A Solid Screw Cap without a liner and a polypropylene Drip Ring are included. This Bottle is often used with the Mobile Phase Delivery System and is Not "Safety Coated".
This item is not returnable. View return policy
Catalog No. 50-227-0181 Supplier Microsolv Technology Corporation Supplier No. 588072LCS
Some of our Bottles, these Clear, Borosilicate Glass Bottles have a 2L "Total Fill Capacity" and are perfect as HPLC Mobile Phase Reservoirs. The small openings, one liter Capacity and White Enamel Graduations on the outside of the Bottles help you determine the Solvent use and prevent running dry. This Bottle can be used with MicroSolv Filter Degassers , other Degassers or disposable Filter Degassers. A Solid Screw Cap without a liner and a polypropylene Drip Ring are included. This Bottle is often used with the Mobile Phase Delivery System and is Not "Safety Coated".